Ethyl 5-amino-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate 98%

Supplier: Ambeed
Warning

A628701-100MG A628701-1G A628701-250MG
77266-938EA 28 USD
77266-938 77266-940 77266-942
Ethyl 5-amino-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate 98%
Ethyl 5-amino-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈F₃N₃O₂
MDL Number: MFCD30011942
CAS Number: 133922-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR