4-(2-(Dimethylamino)ethoxy)benzoic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A627942-100MG
A627942-1G
A627942-250MG
77266-932EA
53
USD
77266-932
77266-934
77266-936
4-(2-(Dimethylamino)ethoxy)benzoic acid hydrochloride 97%
4-(2-(Dimethylamino)ethoxy)benzoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₆ClNO₃ |
MDL Number:
MFCD18252805 CAS Number: 27237-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...