3-Formyl-1H-indole-4-carbonitrile 98%

Supplier: Ambeed
Warning

A627822-100MG A627822-1G A627822-250MG
77242-850EA 28.84 USD
77242-850 77242-852 77242-854
3-Formyl-1H-indole-4-carbonitrile 98%
3-Formyl-1H-indole-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O
MW: 170.16 g/mol
MDL Number: MFCD06657154
CAS Number: 53269-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR