Methanesulfonato(2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct 98%

Supplier: Ambeed
Warning

A627722-100MG A627722-1G A627722-250MG A627722-5G
77254-480EA 8 USD
77254-480 77254-482 77254-484 77573-038
Methanesulfonato(2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct 98%
Methanesulfonato(2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct

Precaución: for Research Use Only

Formula: C₄₃H₆₀NO₃PPdS
MW: 808.4 g/mol
MDL Number: MFCD28411856
CAS Number: 1599466-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR