SH-4-54 ≥98%

Supplier: Ambeed
Warning

Synonyms: SH 4 54, 4-[(4-cyclohexylphenyl)methyl-[2-[methyl-(2,3,4,5,6-pentafluorophenyl)sulphonylamino]acetyl]amino]benzoic acid, SH-454, SH454, SH-4-54, 4-(N-(4-Cyclohexylbenzyl)-2-((2,3,4,5,6-pentafluoro-N-methylphenyl)sulphonamido)acetamido)benzoic acid, SH4-54

A627552-10MG A627552-1MG A627552-25MG A627552-2MG A627552-5MG
77242-840EA 69.01 USD
77242-840 77242-842 77242-844 77242-846 77242-848
SH-4-54 ≥98%
SH-4-54

Precaución: for Research Use Only

Formula: C₂₉H₂₇F₅N₂O₅S
MW: 610.59 g/mol
MDL Number: MFCD28143917
CAS Number: 1456632-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR