(4-Isobutoxyphenyl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A627275-1G A627275-25G A627275-5G
77280-382EA 11.33 USD
77280-382 77280-384 77280-386
(4-Isobutoxyphenyl)methanamine hydrochloride 97%
(4-Isobutoxyphenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₈ClNO
MW: 215.71 g/mol
MDL Number: MFCD16605992
CAS Number: 37806-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR