10-(4-Bromophenyl)-10H-phenoxazine 97%

Supplier: Ambeed
Warning

A627000-1G A627000-250MG
77254-466EA 78 USD
77254-466 77254-468
10-(4-Bromophenyl)-10H-phenoxazine 97%
10-(4-Bromophenyl)-10H-phenoxazine

Precaución: for Research Use Only

Formula: C₁₈H₁₂BrNO
CAS Number: 71041-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR