Methyl-2-chloro-4-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-nitrobenzoic acid methyl ester, Methyl 2-chloro-4-nitrobenzoate

A626826-100G A626826-25G A626826-5G
77242-834EA 350.01 USD
77242-834 77242-836 77242-838
Methyl-2-chloro-4-nitrobenzoate 98%
Methyl-2-chloro-4-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄
MDL Number: MFCD00017013
CAS Number: 13324-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR