3,6-Dioxa-1,8-octanedithiol 97%

Supplier: Ambeed
Danger

Synonyms: Ethylene Glycol Bis(2-mercaptoethyl) Ether, 1,8-Dimercapto-3,6-dioxaoctane, 1,2-Bis(2-mercaptoethoxy)ethane, 2,2′-(Ethylenedioxy)diethanethiol

A625945-25G A625945-100G
77280-380EA 6 USD
77280-380 77551-874
3,6-Dioxa-1,8-octanedithiol 97%
3,6-Dioxa-1,8-octanedithiol

Precaución: for Research Use Only

Formula: C₆H₁₄O₂S₂
MW: 182.30 g/mol
MDL Number: MFCD00015873
CAS Number: 14970-87-7
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR