Bis(4-(3-aminophenoxy)phenyl)methanone 98%

Supplier: Ambeed
Warning

A625679-100G A625679-25G A625679-5G
77266-924EA 86.52 USD
77266-924 77266-926 77266-928
Bis(4-(3-aminophenoxy)phenyl)methanone 98%
Bis(4-(3-aminophenoxy)phenyl)methanone

Precaución: for Research Use Only

Formula: C₂₅H₂₀N₂O₃
MW: 396.44 g/mol
MDL Number: MFCD09908245
CAS Number: 63948-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR