3,5-Dichlorothiophenol ≥95%

Supplier: Ambeed
Danger

A625179-1G A625179-250MG
77280-372EA 35 USD
77280-372 77280-374
3,5-Dichlorothiophenol ≥95%
3,5-Dichlorothiophenol

Precaución: for Research Use Only

Formula: C₆H₄Cl₂S
MDL Number: MFCD00041423
CAS Number: 17231-94-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR