Methyl 2-(2-methoxypyridin-4-yl)acetate 97%

Supplier: Ambeed
Warning

A625115-250MG A625115-1G
77266-922EA 49.44 USD
77266-922 77749-274
Methyl 2-(2-methoxypyridin-4-yl)acetate 97%
Methyl 2-(2-methoxypyridin-4-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD22572382
CAS Number: 464152-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR