2-Methoxy-4-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A624082-10G A624082-250MG A624082-5G
77266-904EA 155 USD
77266-904 77266-906 77266-908
2-Methoxy-4-methylbenzonitrile 98%
2-Methoxy-4-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD11847156
CAS Number: 53078-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR