Fmoc-Orn-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Orn-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-aminopentanoic acid
A623803-1G
A623803-250MG
A623803-5G
77266-894EA
58.71
USD
77266-894
77266-896
77266-898
Fmoc-Orn-OH 97%
Fmoc-Orn-OH
Precaución: for Research Use Only
Formula:
C₂₀H₂₂N₂O₄ MW: 354.40 g/mol |
MDL Number:
MFCD02682439 CAS Number: 158599-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...