3-Chloro-2-fluorocinnamic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(3-Chloro-2-fluorophenyl)acrylic acid

A623697-10G A623697-1G A623697-250MG A623697-5G
77280-364EA 224.54 USD
77280-364 77280-366 77280-368 77280-370
3-Chloro-2-fluorocinnamic acid 95%
3-Chloro-2-fluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆ClFO₂
MW: 200.6 g/mol
MDL Number: MFCD01631392
CAS Number: 261762-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR