N-Methyl-3-(1-(4-(piperazin-1-yl)phenyl)-3-(4'-(trifluoromethyl)-[1,1'-biphenyl]-4-yl)-1H-pyrazol-5-yl)propanamide 98%

Supplier: Ambeed
Warning

Synonyms: OSU-T315

A623292-10MG A623292-1MG A623292-50MG A623292-5MG
77266-886EA 81.37 USD
77266-886 77266-888 77266-890 77266-892
N-Methyl-3-(1-(4-(piperazin-1-yl)phenyl)-3-(4'-(trifluoromethyl)-[1,1'-biphenyl]-4-yl)-1H-pyrazol-5-yl)propanamide 98%
N-Methyl-3-(1-(4-(piperazin-1-yl)phenyl)-3-(4'-(trifluoromethyl)-[1,1'-biphenyl]-4-yl)-1H-pyrazol-5-yl)propanamide

Precaución: for Research Use Only

Formula: C₃₀H₃₀F₃N₅O
MW: 533.58 g/mol
MDL Number: MFCD29924730
CAS Number: 2070015-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR