N-(2-Aminophenyl)-4-(1-((1,3-dimethyl-1H-pyrazol-4-yl)methyl)piperidin-4-yl)benzamide 98+%
Supplier: Ambeed
|
Warning
|
A623282-10MG
A623282-1MG
A623282-25MG
A623282-50MG
A623282-5MG
77266-876EA
42
USD
77266-876
77266-878
77266-880
77266-882
77266-884
N-(2-Aminophenyl)-4-(1-((1,3-dimethyl-1H-pyrazol-4-yl)methyl)piperidin-4-yl)benzamide 98+%
N-(2-Aminophenyl)-4-(1-((1,3-dimethyl-1H-pyrazol-4-yl)methyl)piperidin-4-yl)benzamide
Precaución: for Research Use Only
Formula:
C₂₄H₂₉N₅O |
MDL Number:
MFCD30489755 CAS Number: 934828-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...