(S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%
Supplier: Ambeed
|
Warning
|
A623269-100MG
A623269-1G
A623269-250MG
77254-444EA
98.88
USD
77254-444
77254-446
77254-448
(S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%
(S)-(-)-2,2'-Bis[di(3,5-xylyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₄₆H₄₈O₂P₂ MW: 694.82 g/mol |
MDL Number:
MFCD09753009 CAS Number: 362634-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...