2-Methyl-2-propyl-1,3-propanediol 98%

Supplier: Ambeed
Warning

Synonyms: 2,2-Bis(hydroxymethyl)pentane, 2-Methyl-2-propylpropane-1,3-diol, 2,2-Dimethylolpentane

A623027-1000G A623027-100G A623027-500G
77254-432EA 62.83 USD
77254-432 77254-434 77254-436
2-Methyl-2-propyl-1,3-propanediol 98%
2-Methyl-2-propyl-1,3-propanediol

Precaución: for Research Use Only

Formula: C₇H₁₆O₂
MW: 132.2 g/mol
MDL Number: MFCD00004696
CAS Number: 78-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR