1-Butyl-3-methylimidazolium octylsulphate 99+%

Supplier: Ambeed
Warning

Synonyms: [BMIM] [OctSO4]

A622991-1G A622991-25G A622991-5G
77242-810EA 6 USD
77242-810 77242-812 77242-814
1-Butyl-3-methylimidazolium octylsulphate 99+%
1-Butyl-3-methylimidazolium octylsulphate

Precaución: for Research Use Only

Formula: C₁₆H₃₂N₂O₄S
MDL Number: MFCD03788914
CAS Number: 445473-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR