4-(Difluoromethoxy)pyridin-2-amine 97%

Supplier: Ambeed
Warning

A622757-100MG A622757-250MG A622757-50MG A622757-1G
77266-868EA 44.29 USD
77266-868 77266-870 77266-872 77569-196
4-(Difluoromethoxy)pyridin-2-amine 97%
4-(Difluoromethoxy)pyridin-2-amine

Precaución: For research use only.

Formula: C₆H₆F₂N₂O
MW: 160.121 g/mol
MDL Number: MFCD22550528
CAS Number: 1206972-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR