(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-1-((S)-2-Diphenylphosphino)ferrocenylethylamine, (Rp)-1-[(1S)-(1-Aminoethyl)]-2-(diphenylphosphino)ferrocene

A622355-100MG A622355-1G A622355-250MG A622355-50MG
77254-408EA 176 USD
77254-408 77254-410 77254-412 77254-414
(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine 98%
(S)-1-[(R)-2-(Diphenylphosphino)ferrocenyl]ethylamine

Precaución: for Research Use Only

Formula: C₂₄H₂₄FeNP
MDL Number: MFCD11045439
CAS Number: 607389-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR