2-Mercapto-2-thiazoline 95%

Supplier: Ambeed
Danger

Synonyms: 2-Thiazoline-2-Thiol, 4,5-Dihydrothiazole-2-thiol

A622122-10G A622122-25G
77266-864EA 5.15 USD
77266-864 77266-866
2-Mercapto-2-thiazoline 95%
2-Mercapto-2-thiazoline

Precaución: for Research Use Only

Formula: C₃H₅NS₂
MW: 119.21 g/mol
MDL Number: MFCD00126013
CAS Number: 96-53-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR