Ethyl-4-(Chloromethyl)benzoate 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Chloromethyl)benzoic Acid Ethyl Ester, Ethyl 4-(chloromethyl)benzoate

A621899-1G A621899-25G A621899-5G
77292-928EA 10.3 USD
77292-928 77292-930 77292-932
Ethyl-4-(Chloromethyl)benzoate 98%
Ethyl-4-(Chloromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.65 g/mol
MDL Number: MFCD04337336
CAS Number: 1201-90-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR