4-Trifluoromethoxythioanisole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl(4-(trifluoromethoxy)phenyl)sulphane, 4-(Trifluoromethoxy)phenyl methyl sulphphide, 4-(Trifluoromethoxy)phenyl methyl sulphide
A621875-10G
A621875-5G
77292-924EA
124.63
USD
77292-924
77292-926
4-Trifluoromethoxythioanisole 98%
4-Trifluoromethoxythioanisole
Precaución: for Research Use Only
Formula:
C₈H₇F₃OS MW: 208.20 g/mol |
MDL Number:
MFCD01320776 CAS Number: 2546-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...