Fmoc-Arg(pbf)-H 95%
Supplier: Ambeed
|
Warning
|
A621808-100MG
A621808-1G
A621808-250MG
77242-804EA
135.96
USD
77242-804
77242-806
77242-808
Fmoc-Arg(pbf)-H 95%
Fmoc-Arg(pbf)-H
Precaución: for Research Use Only
Formula:
C₃₄H₄₀N₄O₆S MW: 632.76 g/mol |
MDL Number:
MFCD30476000 CAS Number: 1026023-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...