Chlorobis(ethylene)iridium(I) Dimer 98%

Supplier: Ambeed
Warning

A621532-100MG A621532-1G A621532-250MG
77254-398EA 295 USD
77254-398 77254-400 77254-402
Chlorobis(ethylene)iridium(I) Dimer 98%
Chlorobis(ethylene)iridium(I) Dimer

Precaución: for Research Use Only

Formula: C₈H₁₆Cl₂Ir₂
MDL Number: MFCD28976162
CAS Number: 39722-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR