3-Chloro-4-methylbenzonitrile 97%

Supplier: Ambeed
Danger

A621157-10G A621157-1G A621157-25G A621157-5G
77242-796EA 11 USD
77242-796 77242-798 77242-800 77242-802
3-Chloro-4-methylbenzonitrile 97%
3-Chloro-4-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClN
MDL Number: MFCD00001799
CAS Number: 21423-81-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR