(R)-[2,3':1',1'':3'',2'''-Quaternaphthalene]-2',2''-diol ≥98%, ee 99%

Supplier: Ambeed
Warning

A620758-100MG A620758-1G A620758-250MG
77254-386EA 74 USD
77254-386 77254-388 77254-390
(R)-[2,3':1',1'':3'',2'''-Quaternaphthalene]-2',2''-diol ≥98%, ee 99%
(R)-[2,3':1',1'':3'',2'''-Quaternaphthalene]-2',2''-diol

Precaución: for Research Use Only

Formula: C₄₀H₂₆O₂
MDL Number: MFCD31560044
CAS Number: 215433-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR