3-Methylisothiazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A619276-100MG A619276-1G A619276-250MG A619276-5G
77280-340EA 14 USD
77280-340 77280-342 77280-344 77280-346
3-Methylisothiazole-4-carboxylic acid 95%
3-Methylisothiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₂S
MW: 143.17 g/mol
MDL Number: MFCD00030523
CAS Number: 15903-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR