4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide hydrochloride 97%

Supplier: Ambeed
Warning

A618991-100G A618991-1G A618991-25G A618991-5G
77292-904EA 139.05 USD
77292-904 77292-906 77292-908 77292-910
4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide hydrochloride 97%
4-Amino-1-methyl-3-propyl-1H-pyrazole-5-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅ClN₄O
MW: 218.68 g/mol
MDL Number: MFCD11040573
CAS Number: 247584-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR