3,7-Diaminodibenzo[b,d]thiophene 5,5-dioxide 97%

Supplier: Ambeed
Warning

A618874-100MG A618874-1G A618874-250MG
77242-768EA 18.54 USD
77242-768 77242-770 77242-772
3,7-Diaminodibenzo[b,d]thiophene 5,5-dioxide 97%
3,7-Diaminodibenzo[b,d]thiophene 5,5-dioxide

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂S
MW: 246.28 g/mol
MDL Number: MFCD00159114
CAS Number: 6259-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR