4-Chloro-2-fluoroaniline 98%
Supplier: Ambeed
|
Warning
|
A618085-100G
A618085-10G
A618085-25G
A618085-500G
77292-896EA
34
USD
77292-896
77292-898
77292-900
77292-902
4-Chloro-2-fluoroaniline 98%
4-Chloro-2-fluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₅ClFN |
MDL Number:
MFCD00010625 CAS Number: 57946-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...