3-Methyl-3,9-diazaspiro[5.5]undecane 95%

Supplier: Ambeed
Warning

A617959-1G A617959-100MG A617959-250MG
77280-334EA 399 USD
77280-334 77749-265 77749-266
3-Methyl-3,9-diazaspiro[5.5]undecane 95%
3-Methyl-3,9-diazaspiro[5.5]undecane

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂
MDL Number: MFCD11043237
CAS Number: 13323-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR