Methyl Phenylsulfonylacetate 97%

Supplier: Ambeed
Warning

Synonyms: Phenylsulphonylacetic Acid Methyl Ester

A617892-100G A617892-1G A617892-25G A617892-5G
77254-374EA 197.76 USD
77254-374 77254-376 77254-378 77254-380
Methyl Phenylsulfonylacetate 97%
Methyl Phenylsulfonylacetate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄S
MW: 214.24 g/mol
MDL Number: MFCD00007555
CAS Number: 34097-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR