2-Bromoethyl benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Benzoic Acid 2-Bromoethyl Ester

A617166-1G A617166-5G A617166-25G
77266-838EA 20.6 USD
77266-838 77266-840 77551-848
2-Bromoethyl benzoate 98%
2-Bromoethyl benzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00059462
CAS Number: 939-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR