1-Fluoro-3-ethynylbenzene 96%

Supplier: Ambeed
Danger

Synonyms: (3-Fluorophenyl)ethyne, 3-Fluorophenylacetylene, (3-Fluorophenyl)acetylene, 1-Ethynyl-3-fluorobenzene

A616509-10G A616509-1G A616509-25G A616509-5G
77242-750EA 30.9 USD
77242-750 77242-752 77242-754 77242-756
1-Fluoro-3-ethynylbenzene 96%
1-Fluoro-3-ethynylbenzene

Precaución: for Research Use Only

Formula: C₈H₅F
MW: 120.13 g/mol
MDL Number: MFCD03093798
CAS Number: 2561-17-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR