3-(1H-Pyrazol-3-yl)pyridine 98%

Supplier: Ambeed
Warning

A616421-1G A616421-250MG
77280-330EA 32 USD
77280-330 77280-332
3-(1H-Pyrazol-3-yl)pyridine 98%
3-(1H-Pyrazol-3-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₇N₃
MDL Number: MFCD03407951
CAS Number: 45887-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR