4,4''-Difluoro-5'-(4-fluorophenyl)-1,1':3',1''-terphenyl 98%
Supplier: Ambeed
|
Danger
|
A616256-100MG
A616256-1G
A616256-250MG
A616256-5G
77242-734EA
7
USD
77242-734
77242-736
77242-738
77242-740
4,4''-Difluoro-5'-(4-fluorophenyl)-1,1':3',1''-terphenyl 98%
4,4''-Difluoro-5'-(4-fluorophenyl)-1,1':3',1''-terphenyl
Precaución: for Research Use Only
Formula:
C₂₄H₁₅F₃ |
MDL Number:
MFCD21365089 CAS Number: 448-60-2 UN: 1759 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...