5-Oxo-5-(3-oxopiperazin-1-yl)pentanoic acid 97%

Supplier: Ambeed
Warning

A615902-1G A615902-250MG A615902-5G
77242-728EA 25.75 USD
77242-728 77242-730 77242-732
5-Oxo-5-(3-oxopiperazin-1-yl)pentanoic acid 97%
5-Oxo-5-(3-oxopiperazin-1-yl)pentanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₄
MDL Number: MFCD09045202
CAS Number: 926237-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR