D-Prolinamide 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Pro-NH₂, (R)-2-Pyrrolidinecarboxamide, H-D-Pro-NH2

A615655-100G A615655-1G A615655-25G A615655-5G A615655-500G
77292-878EA 223.51 USD
77292-878 77292-880 77292-882 77292-884 77749-262
D-Prolinamide 97%
D-Prolinamide

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
MDL Number: MFCD00153457
CAS Number: 62937-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR