2-Amino-3',6'-bis(diethylamino)spiro[isoindoline-1,9'-xanthen]-3-one 97%

Supplier: Ambeed
Warning

Synonyms: Rhodamine B Hydrazide

A615462-250MG A615462-100MG A615462-1G A615462-5G
77266-834EA 86.52 USD
77266-834 77617-582 77617-584 77617-586
2-Amino-3',6'-bis(diethylamino)spiro[isoindoline-1,9'-xanthen]-3-one 97%
2-Amino-3',6'-bis(diethylamino)spiro[isoindoline-1,9'-xanthen]-3-one

Precaución: for Research Use Only

Formula: C₂₈H₃₂N₄O₂
MW: 456.57 g/mol
MDL Number: MFCD07808762
CAS Number: 74317-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR