2-Hydroxyanthracene 98%

Supplier: Ambeed
Warning

A614733-1G A614733-250MG A614733-100MG
77266-830EA 96 USD
77266-830 77266-832 77573-002
2-Hydroxyanthracene 98%
2-Hydroxyanthracene

Precaución: for Research Use Only

Formula: C₁₄H₁₀O
MDL Number: MFCD06653605
CAS Number: 613-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR