(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: (R,R)-Me-DUPHOS-Rh

A614441-100MG A614441-1G A614441-250MG
77254-350EA 124 USD
77254-350 77254-352 77254-354
(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate 98%
(-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₂₇H₃₈F₃O₃P₂RhS
MDL Number: MFCD00269862
CAS Number: 187682-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR