2-Benzoylthiophene 98%

Supplier: Ambeed
Warning

A614241-100G A614241-25G A614241-5G
77266-824EA 160 USD
77266-824 77266-826 77266-828
2-Benzoylthiophene 98%
2-Benzoylthiophene

Precaución: for Research Use Only

Formula: C₁₁H₈OS
MDL Number: MFCD00022495
CAS Number: 135-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR