4-Bromo-1,10-phenanthroline 98%

Supplier: Ambeed
Warning

A614235-100MG A614235-250MG
77242-724EA 24 USD
77242-724 77242-726
4-Bromo-1,10-phenanthroline 98%
4-Bromo-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₂H₇BrN₂
MDL Number: MFCD28016364
CAS Number: 7089-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR