1-Bromo-3-fluoro-5-iodobenzene 98%

Supplier: Ambeed
Warning

A613806-100G A613806-10G A613806-1G A613806-250MG A613806-25G
77292-868EA 356 USD
77292-868 77292-870 77292-872 77292-874 77292-876
1-Bromo-3-fluoro-5-iodobenzene 98%
1-Bromo-3-fluoro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
MDL Number: MFCD06657969
CAS Number: 845866-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR