5-Amino-3-(4-bromophenyl)-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Bromophenyl)-1H-pyrazol-5-amine

A613710-100G A613710-1G A613710-25G A613710-5G
77266-816EA 631.39 USD
77266-816 77266-818 77266-820 77266-822
5-Amino-3-(4-bromophenyl)-1H-pyrazole 98%
5-Amino-3-(4-bromophenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₈BrN₃
MW: 238.09 g/mol
MDL Number: MFCD00558911
CAS Number: 78583-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR