4-Bromo-2,3-difluorobenzoic acid 96%

Supplier: Ambeed
Warning

A613318-100MG A613318-10G A613318-1G A613318-250MG A613318-25G A613318-5G A613318-100G
77292-854EA 7 USD
77292-854 77292-856 77292-858 77292-860 77292-862 77292-864 77569-162
4-Bromo-2,3-difluorobenzoic acid 96%
4-Bromo-2,3-difluorobenzoic acid

Precaución: For research use only.

Formula: C₇H₃BrF₂O₂
MDL Number: MFCD07368121
CAS Number: 194804-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR