1-Methylpyrrole 98%

Supplier: Ambeed
Danger

Synonyms: N-Methylpyrrole, 1-Methyl-1H-pyrrole

A613016-1G A613016-25G A613016-5G A613016-10G
77242-710EA 5.15 USD
77242-710 77242-712 77242-714 77747-264
1-Methylpyrrole 98%
1-Methylpyrrole

Precaución: for Research Use Only

Formula: C₅H₇N
MW: 81.12 g/mol
MDL Number: MFCD00005345
CAS Number: 96-54-8
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR